Learn More
Abnova™ Annexin V-Biotin Apoptosis Kit
Annexin V-Biotin Apoptosis Kit is used for apoptosis detection.
Manufacturer:  Abnova™ KA0709
Catalog No. 89-027-065
Add to cart
Description
Description
  • Kit is based on the observation that soon after initiating apoptosis, most cell types translocate the membrane phospholipid phosphatidylserine (PS) from the inner face of the plasma membrane to the cell surface
  • Can be detected in conjunction with conventional dye-staining using any streptavidine- or avidin-dye reagents, such as (strept) avidine-fluorescein, -peroxidase, -alkaline phosphatase (AP), and –ß-gal, etc.
  • Store kit at 4°C, stable for one year
Specifications
Specifications
Annexin V-Biotin Apoptosis Kit is used for apoptosis detection.
Biotin
1 Year
1 Kit
Flow Cytometry, Functional Study
Store the kit at 4°C.