Learn More
Abnova™ Annexin V-Cy5 Apoptosis Kit
Annexin V-Cy5 Apoptosis Kit is used for apoptosis detection.
Manufacturer:  Abnova™ KA0712
Catalog No. 89-027-068
Add to cart
Description
Description
  • Kit is based on the observation that soon after initiating apoptosis, cells translocate the membrane phosphatidyl-serine (PS) from the inner face of the plasma membrane to the cell surface
  • Once on the surface, PS can be easily detected by staining with a fluorescent conjugate of Annexin V, a protein that has a high affinity for PS
  • One-step staining procedure takes only 10 minutes
  • Detection can be analyzed by flow cytometry or by fluorescence microscopy
  • Store kit at 4°C
Specifications
Specifications
Annexin V-Cy5 Apoptosis Kit is used for apoptosis detection.
Fluorescent
Store the kit at 4°C.
1 Kit
Flow Cytometry, Functional Study