Learn More
Abnova™ Annexin V-EGFP Apoptosis Kit
Annexin V-EGFP Apoptosis Kit is used for apoptosis detection.
Manufacturer:  Abnova™ KA0713
Catalog No. 89-027-069
Add to cart
Description
Description
  • Kit is based on the observation that soon after initiating apoptosis, most cell types translocate the membrane phospholipid phosphatidylserine (PS) from the inner face of the plasma membrane to the cell surface
  • Once on the surface, PS can be easily detected by staining with an enhanced green fluorescent protein (EGFP) fusion of annexin V, a protein that has a strong natural affinity for PS
  • One-step staining procedure takes only 10 minutes
  • Assay can be directly performed on live cells
  • Detection can be analyzed by flow cytometry or by fluorescent reagents
  • Kit can differentiate apoptosis vs. necrosis when performing both annexin V-EGFP and PI staining
  • Store kit at 4°C; stable for one year
Specifications
Specifications
Annexin V-EGFP Apoptosis Kit is used for apoptosis detection.
Flow Cytometry, Functional Study
Store the kit at 4°C.
1 Kit
1 Year