Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Annexin V-FITC Apoptosis Kit (Premium)

Used for apoptosis detection

Manufacturer:  Abnova™ KA0715

Catalog No. 89-027-071


Add to cart

Description

Description

Annexin V-FITC Apoptosis Kit (Premium) using SYTXO green dye and annexin V-FITC for apoptosis detection

  • Sample Type: Cell
  • Species Cross reactivity: Mammals

Functional Study; Flow Cytometry

Specifications

Specifications

Annexin V-FITC Apoptosis Kit (Premium) using SYTXO green dye and annexin V-FITC for apoptosis detection.
Fluorescent
1 Year
1 Kit
Flow Cytometry, Functional Study
Store the kit at 4°C.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit