Learn More
Abnova™ Annexin V-FITC Apoptosis Kit
Annexin V-FITC Apoptosis Kit is used for apoptosis detection.
Manufacturer:  Abnova™ KA0714
Catalog No. 89-027-070
Add to cart
Description
Description
  • Kit is based on the observation that soon after initiating apoptosis, cells translocate the membrane phosphatidylserine (PS) from the inner face of the plasma membrane to the cell surface
  • Once on the surface, PS can be easily detected by staining with a fluorescent conjugate of Annexin V, a protein that has a high affinity for PS
  • One-step staining procedure takes only 10 minutes
  • Detection can be analyzed by flow cytometry or by fluorescence microscopy
  • Kit can differentiate apoptosis and necrosis when performing both Annexin V-FITC and PI staining
  • Store kit at 4°C; stable for one year
Specifications
Specifications
Annexin V-FITC Apoptosis Kit is used for apoptosis detection.
Fluorescent
1 Year
1 Kit
Flow Cytometry, Functional Study
Store the kit at 4°C.