Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Antibody Purification Kit

For measuring Trypsin activity using fluorometric methods

Manufacturer:  Abnova™ KA1595

Catalog No. 89-028-278


Add to cart

Specifications

Specifications

The Antibody Purification Kit quickly removes the contaminants which interfere in labeling reactions; it can also be used to purify antibodies from crude samples such as ascites fluid or immune serum.
Store at 4°C.
1 Kit
Purification
Antibody Purification Kit
1 Vial lyophilized Protein A resin, 1 vial 10X Binding Buffer, 1 vial Wash Buffer, 1 vial Elution Buffer, 1 vial neutraliser
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit