Abnova™ APOA2 (Human) ELISA Kit Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: Abnova Corporation KA0461

 View more versions of this product

Catalog No. 89-022-888

  Check Availability
Add to Cart

    Specifications

    Cross Reactivity Human
    Detection Method Colorimetric
    Product Type APOA2 (Human) ELISA kit
    Quantity 96 Assays
    Sample Type Cell Culture Supernatant, Milk, Plasma, Serum, Urine
    Volume (Metric) Sample 50μL
    Conjugate Biotin
    For Use With (Application) Quantification, for the quantitative measurement of Human APOA2
    Includes 96-well Antibody coated microwell, 8μg Human apo A-II standard , 1 vial Biotinylated apo A-II Antibody (100X), 30mL EIA diluent concentrate (10X), 30mL x 2 Wash Buffer 20X, 80 Streptavidin-Peroxidase Conjugate (SP Conjugate), 8mL chromog
    Storage Requirements 4°C

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit