Learn More
Abnova™ Apoptotic Cell Isolation Kit
Apoptotic Cell Isolation Kit is used for apoptotic cells isolation or removal of dead cells from cell culture or tissue preparations using annexin V/magnetic beads.
Manufacturer:  Abnova™ KA0719
Catalog No. 89-027-075
Add to cart
Description
Description
  • Provides a simple and efficient means for isolation of apoptotic cells or removal of dead cells from cell culture or tissue preparations using Annexin V/magnetic beads
  • Separated apoptotic and healthy cells can be used in a variety of assays to study apoptotic mechanisms and pathways
  • Kit has also been successfully used to remove dead cells from healthy cell culture
  • Configured for research use only and is not for diagnostic and clinical use
  • For short term, store all components at 4°C; for long term, store at -20°C
Specifications
Specifications
Apoptotic Cell Isolation Kit is used for apoptotic cells isolation or removal of dead cells from cell culture or tissue preparations using annexin V/magnetic beads.
Fluorescent
Store the kit at -20°C.
1 Kit
Functional Study