Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Apoptotic DNA Ladder Detection Kit (Premium)

Apoptotic DNA Ladder Detection Kit (Premium) is used for detecting DNA fragmentation in apoptotic cells with a high sensitive dye.

Manufacturer:  Abnova™ KA0721

Catalog No. 89-027-077


Add to cart

Includes:
  • TE lysis buffer, 1.8mL
  • Enzyme B (lyophilized), 1 vial
  • Ammonium acetate solution, 0.25mL
  • DNA suspension buffer, 0.25mL
  • Staining dye (10000X), 50µL

Description

Description

  • Provides an easy and sensitive means for detecting DNA fragmentation in apoptetic cells
  • New detection method requires less than 90 minutes to prepare DNA, with neither extraction nor using columns
  • Fragmentation can be easily visualized by agarose gel electrophoreses stained with a highly sensitive dye
  • New procedure provides higher sensitivity in comparison to other similar kits in the market
  • Assay can be used to detect apoptotic DNA ladder in both tissues and cells
  • Store kit at -20°C

Specifications

Specifications

Apoptotic DNA Ladder Detection Kit (Premium) is used for detecting DNA fragmentation in apoptotic cells with a high sensitive dye.
1 Kit
Functional Study
Store the kit at -20°C.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit