Learn More
Abnova™ Apoptotic DNA Ladder Detection Kit
Apoptotic DNA Ladder Detection Kit is used for detecting DNA fragmentation in apoptotic cells.
Manufacturer:  Abnova™ KA0720
Catalog No. 89-027-076
Add to cart
Description
Description
  • Provides an easy and sensitive means for detecting DNA fragmentation in apoptetic cells
  • New detection method requires less than 90 minutes to prepare DNA, with neither extraction nor using columns
  • Fragmentation can be easily visualized by agarose gel electrophoreses
  • New procedure increases recovery of small fragmented DNA and therefore improves the sensitivity of the assay
  • Store kit at -20°C
Specifications
Specifications
Apoptotic DNA Ladder Detection Kit is used for detecting DNA fragmentation in apoptotic cells.
Liquid (NP-40), Liquid (Glycerol), Powder (Ammonium Acetate)
Store the kit at -20°C.
1 Kit
Functional Study