Learn More
Abnova™ Apoptotic DNA Ladder Isolation Kit
Provides an easy and sensitive means for detecting DNA fragmentation in apoptotic cells. The extracted apoptotic DNA ladder fragments can be easily visualized by agarose gel electrophoreses.
Manufacturer:  Abnova™ KA0722
Catalog No. 89-027-078
Add to cart
Description
Description
Apoptotic DNA Ladder Isolation Kit is used for extract apoptotic DNA ladder from both tissues and cells with as little as 5% or less apoptotic cell population
Functional Study
Specifications
Specifications
Apoptotic DNA Ladder Isolation Kit is used for extract apoptotic DNA ladder from both tissues and cells with as little as 5% or less apoptotic cell population.
Liquid (NP-40), Liquid (Glycerol), Solid (Ammonium Acetate)
Store the kit at -20°C.
1 Kit
Functional Study