Learn More
Abnova™ Abnova™ Aspartate Assay Kit (Colorimetric/Fluorometric)
Manufacturer:  Abnova™ KA3708
 View more versions of this product
Catalog No. 89-101-227
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Colorimetric: 0.1 to 10nM, Fluorometric: 0 to 25nM
Biological Fluid, Cell Culture Supernatant, Culture Medium, Plasma, Serum, Tissue Culture Supernatant, Urine
Colorimetric, Fluorometric
For quantitative colorimetric/fluorometric determination of aspartate
Not Applicable