Learn More
Abnova™ Abnova™ Aspartate Assay Kit (Colorimetric/Fluorometric)
Manufacturer:  Abnova™ KA3790
 View more versions of this product
Catalog No. 89-101-206
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Colorimetric, Fluorometric
For quantitative colorimetric/fluorometric determination of aspartate at OD570, FL530/585nm
25μL
Colorimetric: 2 to 400μM, Fluorometric: 1 to 50μM
Culture Medium, Plasma, Serum, Tissue