Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ATP Assay Kit (Colorimetric/Fluorometric)

To detect ATP, functional study

Manufacturer:  Abnova™ KA0806

Catalog No. 89-028-092


Add to cart

Includes:
  • ATP assay buffer, 25mL
  • ATP probe (in DMSO), 0.2mL
  • ATP converter, 1 vial
  • Developer mix (lyophilized), 1 vial
  • ATP standard (1µmol, lyophilized), 1 vial

Specifications

Specifications

ATP Assay Kit (Colorimetric/Fluorometric) is used for detecting ATP.
25mL ATP assay buffer, 0.2mL ATP probe (in DMSO), 1 vial ATP converter, 1 vial developer mix (lyophilized), 1 vial ATP standard (1μM) (lyophilized)
Biological fluid, cell lysate, culture medium, plasma, serum, tissue lysate, urine
Colorimetric, Fluorometric
Functional Study
1 Kit
Store the kit at -20°C.
ATP Assay Kit
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit