Learn More
Abnova™ ATP Cell Viability Assay Kit
To detect the ATP levels for a rapid screening of apoptosis and cell proliferation simultaneously in mammalian cells, functional study
Manufacturer:  Abnova™ KA0794
Catalog No. 89-028-100
Add to cart
Specifications
Specifications
ATP Cell Viability Assay Kit is used for detecting the ATP levels for a rapid screening of apoptosis and cell proliferation simultaneously in mammalian cells.
20mL Nucleotide Releasing buffer, 1 vial ATP Monitoring enzyme, 2.15mL enzyme Reconstitution buffer, 1mg ATP
Biological fluid, cell lysate, culture medium, plasma, serum, soil, sludge, tissue lysate, urine
ATP Cell Viability Assay Kit
Functional Study
1 Kit
Store the kit at -20°C.