Learn More
Abnova™ beta-Hydroxybutyrate Assay Kit
Used for measuring beta-HB levels and functional study
Manufacturer:  Abnova™ KA0854
Catalog No. 89-024-409
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
0.01 to 0.2mm
Functional Study
1 Kit
Store the kit at -20°C.
beta-Hydroxybutyrate Assay Kit
beta-Hydroxybutyrate Assay Kit is used for measuring beta-HB levels.
25mL beta-HB assay buffer, 1 vial beta-HB enzyme mix (lyophilized), 1 vial beta-HB substrate mix (lyophilized), 1 vial beta-HB standard (1μM, lyophilized)
Biological fluid, cell culture supernatant, plasma, serum, tissue culture supernatant, urine
Colorimetric