Learn More
Abnova™ Bioluminescence Cytotoxicity Assay Kit
Used for detecting cell death or cytotoxicity and viability and functional study
Manufacturer:  Abnova™ KA0898
Catalog No. 89-024-447
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
Bioluminescence Cytotoxicity Assay Kit is used for detecing cell death or cytotoxicity.
5 vials AK detection reagent (lyophilized), 50mL AK assay buffer
Cell
Bioluminescence Cytotoxicity Assay Kit
Functional Study
1 Kit
Store the kit at -20°C.