Learn More
Abnova™ Blastocystis spp. ELISA Kit
ELISA for the detection of Blastocystis species antigen in Human stool
Manufacturer:  Abnova™ KA2092
Catalog No. 89-028-823
Add to cart
Specifications
Specifications
HRP
Blastocystis spp. ELISA Kit is an enzyme linked immunosorbent assay for the detection of Blastocystis spp. antigen in human stool.
Quantification
Blastocystis spp. ELISA Kit
Stool
400μL
Human
Colorimetric
Microtiter plate coated with Anti-Blastocystis Polyclonal Antibodies 96 break apart well (8x12), 1 bottle, 100mL 20X concentrated Wash Buffer,1 bottle 60mL/2 bottles 50mL Stool Diluent (Blue), 1 bottle 12mL/1 bottle 16mL HRP Conjugate (Green), 2mL/1 vial
96 Assays
Store the kit at 4°C. Do not freeze.