Learn More
Abnova™ BLUE Phosphatase Substrate Kit
Immunohistochemistry
Manufacturer:  Abnova™ KA0567
Catalog No. 89-027-131
Add to cart
Specifications
Specifications
BLUE Phosphatase Substrate Kit contains reagents for the detection of alkaline phosphatase-labeled conjugates. BLUE reagents form a royal blue reaction product that is alcohol soluble, and aqueous mounting media should be used. Contrast RED, a nuclear counterstain formulated from nuclear Fast Red, provides excellent nuclear detail.
10mL Blue solution, 10mL Activator solution, 50mL Buffered Substrate solution, 50mL Contrast Red solution
Blue Phosphatase Substrate kit
Store the kit at 4°C.
Immunohistochemistry
BLUE Phosphatase Substrate Kit
1 Kit