Learn More
Abnova™ Borrelia burgdorferi IgM ELISA Kit
For the detection and the quantitative determination of specific IgM antibodies against Borrelia burgdorferi in Serum and Plasma
Manufacturer:  Abnova™ KA1283
Catalog No. 89-024-575
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
HRP
Borrelia burgdorferi IgM Ab ELISA Kit has been designed for the detection and the quantitative determination of specific IgM antibodies against Borrelia burgdorferi in serum and plasma.
Quantification
Borrelia burgdorferi IgM ELISA Kit
Plasma, Serum
100μL
Human
Colorimetric
Borrelia burgdorferi Antigen coated microtiter strip 12, 2mL calibrator A (negative control), 2mL calibrator B (cut-Off standard), 2mL calibrator C (weak positive control), 2mL calibrator D (positive control),15mL Enzyme Conjugate, 15mL Substrate, 15mL stop solution, 60mL 10X sample diluent 60mL, washing Buffer
1 Kit
Store the kit at 4°C.