Learn More
Abnova™ BPI Ab ELISA Kit
ELISA, Immunohistochemistry, In-situ Hybridization, Western Blotting
Manufacturer:  Abnova™ KA1088
Catalog No. 89-027-226
Add to cart
Specifications
Specifications
6.3 to 100U/mL
BPI Ab ELISA Kit is an indirect solid phase ELISA for the quantitative measurement of IgG class autoantibodies against BPI in human serum or plasma.
Quantification
BPI Ab ELISA Kit
Plasma, Serum
10μL
HRP
Colorimetric
Combined calibrator with IgG, 1.5mL each class Anti-BPI 6 vials, 1.5mL each Anti-BPI controls 2 vials, 20mL Sample Buffer 1 vial,15mL Enzyme Conjugate solution 1 vial, 15mL TMB Substrate solution 1 vial, 15mL stop solution 1 vial, 96-well plate 1 plate, 20mL wash solution 1 vial
96 Tests
Store the kit at 4°C.