Learn More
Abnova™ Branched Chain Amino Acid (Leu, Ile, Val) Assay Kit
To measure the branched chain amino acid (Leu, Ile, Val), functional study
Manufacturer:  Abnova™ KA0824
Catalog No. 89-028-075
Add to cart
Specifications
Specifications
0 to 10nm
Functional Study
1 Kit
Store the kit at -20°C.
Branched Chain Amino Acid (Leu, Ile, Val) Assay Kit
Branched Chain Amino Acid (Leu, Ile, Val) Assay Kit is used for measuring the BCAA
25mL BCAA assay buffer, 1 vial BCAA enzyme mix, 1 vial WST substrate mix, 100μL Leu standard
Biological fluid, cell culture supernatant, plasma, serum, tissue culture supernatant, urine
Colorimetric