Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ BSG (Human) ELISA Kit

Sandwich Enzyme Immunoassay for the quantitative measurement of BSG

Manufacturer:  Abnova™ KA1006

Catalog No. 89-028-132


Add to cart

Specifications

Specifications

0.78 to 50ng/mL
Human
0.3ng/mL
Quantification
BSG (Human) ELISA Kit
1 Kit
Store the kit at -20°C.
HRP
BSG (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of BSG.
Colorimetric
Antibody pre coated 96-well microplate 1 plate 6 strip, 120μL 100X Biotinylated antibody 1 vial, 120μL 100X HRP Conjugated Streptavidin 1 vial, 30mL Antibody Diluents 1 vial, wash 30mL (20 x) solution concentrated 1 via
BSG (Human) ELISA kit
Cell Culture Supernatant, Urine
50μL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit