Learn More
Abnova™ cAMP Direct Immunoassay Kit
cAMP Direct Immunoassay Kit is used for determining cAMP level.
Manufacturer:  Abnova™ KA0886
Catalog No. 89-024-441
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Kit provides a direct competitive immunoassay for sensitive and quantitative determination of cAMP level in biological samples
  • Kit utilizes recombinant Protein G coated 96-well plate to efficiently anchor cAMP polyclonal antibody onto the plate
  • Kit provides a new acetylation procedure to improve detection sensitivity significantly
  • Kit can detect approximately 0.1 to 10 pmol/well cAMP samples
  • Kit should be stored at -20°C; after opening and reconstitution, components can be stored as instructed, stable for 1 to 2 months
Specifications
Specifications
1 Kit
Store the kit at -20°C.
Functional Study
cAMP Direct Immunoassay Kit is used for determining cAMP level.
Colorimetric