Learn More
Abnova™ Cannabinoids ELISA Kit
Specific and sensitive in-vitro test to detect the presence of Cannabinoids in samples such as whole Blood, Serum, Plasma and Urine
Manufacturer:  Abnova™ KA0928
Catalog No. 89-024-469
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
2 to 10ng/mL
Cannabinoids ELISA Kit is a specific and sensitive in-vitro test to detect the presence of cannabinoids in samples such as whole blood, serum, plasma and urine.
Colorimetric
Microwell coated with Polyclonal anti carboxy THC 96 (8 x 12) well, 12mL THC Conjugate, 2mL positive Refrence standard, 1mL negative standard, 12mL TMB Substrate, 11mL stop solution
96 Assays
Store the kit at 4°C.
HRP
1ng/mL
Qualitative
Cannabinoids ELISA Kit
Plasma, Serum, Urine, Whole Blood
10μL