Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Caspase-1 Inhibitor Screening Kit

Caspase-1 Inhibitor Screening Kit is used for screening caspase inhibitors using fluorometric methods.

Manufacturer:  Abnova™ KA0729

Catalog No. 89-027-085


Add to cart

Description

Description

  • Kit provides an effective means for screening caspase inhibitors using fluorometric methods
  • Simple, straightforward, and can be performed directly in microtiter plates
  • Can be quantified by fluorometry
  • Assay conditions have been optimized to obtain the maximal activity
  • Kit has been configured for research only and is not for diagnostic and clinical use
  • Store kit at -20°C (store 2X Reaction Buffer at 4°C after opening
  • All reagents are stable for 6 months under proper storage conditions

Specifications

Specifications

Caspase-1 Inhibitor Screening Kit is used for screening caspase inhibitors using fluorometric methods.
Liquid (DMSO), Powder (DTT)
Functional Study
1 Kit
Fluorometric
Store the kit at -20°C.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit