Learn More
Abnova™ Caspase-10 Assay Kit (Fluorometric)
Caspase-10 Assay Kit (Fluorometric) is used for detecting the activity of caspases that recognize the sequence AEVD using fluorometric method.
Manufacturer:  Abnova™ KA0731
Catalog No. 89-027-087
Add to cart
Description
Description
  • Kit provides a simple and convenient means for assaying the activity of caspases that recognize the sequence AEVD
  • Assay is based on detection of cleavage of substrate AEVD-AFC
  • Can be quantified using a fluorometer or a fluorescence microtiter plate reader
  • Kit has been configured for research only and is not for diagnostic and clinical use
  • Store kit at -20°C (store Cell Lysis Buffer and 2X Reaction Buffer at 4°C after opening); protect AEVD-AFC from light
  • All reagents are stable for 6 months under proper storage conditions
Specifications
Specifications
Caspase-10 Assay Kit (Fluorometric) is used for detecting the activity of caspases that recognize the sequence AEVD using fluorometric method.
Liquid (DMSO), Powder (DTT)
Functional Study
1 Kit
Colorimetric
Store the kit at -20°C.