Learn More
Abnova™ Caspase-10 Inhibitor Screening Kit
Caspase-10 Inhibitor Screening Kit is used for screening caspase inhibitors using fluorometric methods.
Manufacturer:  Abnova™ KA0732
Catalog No. 89-027-088
Add to cart
Description
Description
  • Kit provides an effective means for screening caspase inhibitors using fluorometric methods
  • Assay utilizes synthetic peptide substrate AEVD-AFC
  • Level of inhibition of caspase-10 activity can be determined by comparison of the fluorescence intensity in samples with and without the testing inhibitors
  • Simple, straightforward, and can be performed directly in microtiter plates
  • Assay conditions have been optimized to obtain the maximal activity
  • Store kit at -20°C
Specifications
Specifications
Caspase-10 Inhibitor Screening Kit is used for screening caspase inhibitors using fluorometric methods.
Liquid (DMSO), Powder (DTT)
Functional Study
1 Kit
Colorimetric
Store the kit at -20°C.