Learn More
Abnova™ Caspase-12 Assay Kit (Fluorometric)
Caspase-12 Assay Kit (Fluorometric) is used for detecting the activity of caspases that recognize the sequence ATAD using fluorometric method.
Manufacturer:  Abnova™ KA0733
Catalog No. 89-027-089
Add to cart
Description
Description
  • Kit provides a simple and convenient means for assaying the activity of caspases that recognize the sequence ATAD
  • Assay is based on detection of cleavage of substrate ATAD-AFC
  • Can be quantified using a fluorometer or a fluorescence microtiter plate reader
  • Store kit at -20°C (store Cell Lysis Buffer and 2X Reaction Buffer at 4°C after opening)
  • All reagents are stable for 6 months from date of receipt under proper storage conditions
Specifications
Specifications
Caspase-12 Assay Kit (Fluorometric) is used for detecting the activity of caspases that recognize the sequence ATAD using fluorometric method.
Liquid (DMSO), Powder (DTT)
FITC-ATAD-FMK
Store the kit at -20°C.
1 Kit
Colorimetric
Functional Study