Learn More
Abnova™ Caspase-2 Assay Kit (Fluorometric)
Caspase-2 Assay Kit (Fluorometric) is used for detecting the activity of caspases that recognize the sequence VDVAD using fluorometric method.
Manufacturer:  Abnova™ KA0736
Catalog No. 89-027-092
Add to cart
Description
Description
  • Kit provides a simple and convenient means for assaying the activity of caspases that recognize the sequence VDVAD
  • Assay is based on detection of cleavage of substrate VDVAD-AFC
  • Can be quantified using a fluorometer or fluorescence microtiter plate reader
  • Kit has been configured for research use only and is not for diagnostic and clinical use
  • Store kit at -20°C
Specifications
Specifications
Caspase-2 Assay Kit (Fluorometric) is used for detecting the activity of caspases that recognize the sequence VDVAD using fluorometric method.
Liquid (Triton X-100), Liquid (DMSO), Powder (DTT)
Functional Study
1 Kit
Fluorometric
Store the kit at -20°C.