Learn More
Abnova™ Caspase-3 Assay Kit (Fluorometric) Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Abnova Corporation KA0741
 View more versions of this product
Catalog No. 89-027-097
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
1 Kit
Liquid (Triton X-100), Liquid (Glycerol), Liquid (DMSO), Powder (DTT)
Detecting the activity of Caspases that recognize the sequence DEVD using fluorometric method, functional study
Assay kit
Colorimetric
-20°C