Learn More
Abnova™ Caspase-3 Immunoassay Kit
Caspase-3 Immunoassay Kit is used for specific, quantitative detection of caspase-3 activity.
Manufacturer:  Abnova™ KA0739
Catalog No. 89-027-095
Add to cart
Description
Description
  • Kit provides an effective immunosorbent enzyme assay for specific, quantitative detection of caspase-3 activity in microtiter plates
  • Assay utilizes caspase-3 polyclonal antibody to capture activated caspase-3 from cell lysates
  • Cleavage generates free AFC which can be analyzed fluorometrically using a fluorescence plate reader
  • Assay ensures absolute specific detection of caspase-3
  • Store kit at -20°C
Specifications
Specifications
Caspase-3 Immunoassay Kit is used for specific, quantitative detection of caspase-3 activity.
Liquid (Triton X-100), Liquid (Glycerol), Liquid (DMSO), Powder (DTT)
Functional Study
1 Kit
Fluorometric
Store the kit at -20°C.