Learn More
Abnova™ Caspase-3 Staining Kit (Green)
Caspase-3 Staining Kit (Green) is used for sensitive detection of activated caspase-3 in living cells with fluorescein.
Manufacturer:  Abnova™ KA0743
Catalog No. 89-027-099
Add to cart
Description
Description
  • Kit provides a convenient means for sensitive detection of activated caspase-3 in living cells
  • Assay utilizes the caspase-3 inhibitor, DEVD-FMK, conjugated to FITC as a marker
  • FITC-DEVD-FMK is cell permeable, nontoxic, and irreversibly binds to activated caspase-3 in apoptotic cells
  • FITC label allows for direct detection of activated caspases in apoptotic cells by fluorescence microscopy, flow cytometry, or fluorescence plate reader
  • Store kit at -20°C
Specifications
Specifications
Caspase-3 Staining Kit (Green) is used for sensitive detection of activated caspase-3 in living cells with fluorescein.
Liquid (DMSO)
FITC-DEVD-FMK
Store the kit at -20°C.
1 Kit
Fluorometric
Flow Cytometry, Functional Study