Learn More
Abnova™ Caspase-3 Staining Kit (Red)
Caspase-3 Staining Kit (Red) is used for sensitive detection of activated caspase-3 in living cells with sulfo-rhodamine.
Manufacturer:  Abnova™ KA0744
Catalog No. 89-028-036
Add to cart
Description
Description
  • Kit provides a convenient means for detecting activated caspase-3 in living cells
  • Assay utilizes the caspase-3 inhibitor, DEVD-FMK, conjugated to sulfo-rhodamine (Red-DEVD-FMK) as the fluorescent in situ marker
  • Red-DEVD-FMK is cell permeable, nontoxic, and irreversibly binds to activated caspase-3 in apoptotic cells
  • Red fluorescence label allows for direct detection of activated caspase-3 in apoptotic cells by fluorescence microscopy, flow cytometry, or fluorescence plate reader
  • Store kit at -20°C
Specifications
Specifications
Caspase-3 Staining Kit (Red) is used for sensitive detection of activated caspase-3 in living cells with sulfo-rhodamine.
Liquid (DMSO)
FITC-DEVD-FMK
Store the kit at -20°C.
1 Kit
Fluorometric
Flow Cytometry, Functional Study