Learn More
Abnova™ Caspase-5 Inhibitor Screening Kit
Caspase-5 Inhibitor Screening Kit is used for screening caspase inhibitors using fluorometric methods.
Supplier:  Abnova™ KA0750
Catalog No. 89-028-030
Add to Cart
Description
Description
  • Kit provides an effective means for screening caspase inhibitors using fluorometric methods
  • Assay utilizes synthetic peptide substrate WEHD-AFC
  • Free AFC can be quantified using fluorometry
  • Simple, straightforward, and can be performed directly in microtiter plates
  • Assay conditions have been optimized to obtain the maximal activity
  • Kit has been configured for research use only and is not for diagnostic and clinical use
  • Store kit at -20°C (store 2X Reaction Buffer at 4°C after opening)
  • All reagents are stable for 6 months under proper storage conditions
Specifications
Specifications
Caspase-5 Inhibitor Screening Kit is used for screening caspase inhibitors using fluorometric methods.
Store the kit at -20°C
Liquid (DMSO), Powder (DTT)
1 Kit
Functional Study
Product Suggestions
Product Suggestions