Learn More
Abnova™ Caspase-8 Staining Kit (Red)
Caspase-8 Staining Kit (Red) is used for sensitive detection of activated caspase-8 in living cells with sulfo-rhodamine.
Manufacturer:  Abnova™ KA0760
Catalog No. 89-028-020
Add to cart
Description
Description
  • Kit provides a convenient means for detecting activated caspase-8 in living cells
  • Assay utilizes the caspase-8 inhibitor, IETD-FMK, conjugated to sulfo-rhodamine (Red-IETD-FMK) as the fluorescent marker
  • Red-IETD-FMK is cell permeable, nontoxic, and irreversibly binds to activated caspase-8 in apoptotic cells
  • The fluorescence label allows detection of activated caspase-8 in apoptotic cells directly by fluorescence microscopy, flow cytometry, or fluorescence plate reader
  • Kit has been configured for research use only and is not for diagnostic and clinical use
  • Store kit at -20°C
Specifications
Specifications
Caspase-8 Staining Kit (Red) is used for sensitive detection of activated caspase-8 in living cells with sulfo-rhodamine.
Liquid (DMSO)
FITC-IETD-FMK
Store the kit at -20°C.
1 Kit
Fluorometric
Flow Cytometry, Functional Study