Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Caspase-9 Assay Kit (Colorimetric)

Caspase-9 Assay Kit (Colorimetric) is used for detecting the activity of caspases that recognize the sequence LEHD using colorimetric method.

Manufacturer:  Abnova™ KA0761

Catalog No. 89-028-019


Add to cart

Description

Description

  • Kit provides a simple and convenient means for assaying the activity of caspases that recognize the sequence LEHD
  • Assay is based on spectrophotometric detection of the chromophore p-nitroanilide (pNA) after cleavage from the labeled substrate LEHD-pNA
  • pNA light emission can be quantified using a spectrophotometer or a microtiter plate reader at 400- or 405-nm
  • Store kit at -20°C (store Cell Lysis Buffer, 2X Reaction Buffer, and Dilution Buffer at 4°C after opening)
  • All reagents are stable for at least 6 months under proper storage conditions

Specifications

Specifications

Caspase-9 Assay Kit (Colorimetric) is used for detecting the activity of caspases that recognize the sequence LEHD using colorimetric method.
Liquid (Triton X-100), Liquid (DMSO), Liquid (Glycerol), Powder (DTT)
Functional Study
1 Kit
Colorimetric
Store the kit at -20°C.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit