Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Abnova™ Caspase-9 Staining Kit (Green)
Caspase-9 Staining Kit (Green) is used for sensitive detection of activated caspase-9 in living cells with fluorescein.
Manufacturer:  Abnova™ KA0764
Catalog No. 89-028-016
Description
Description
  • Kit provides a convenient means for sensitive detection of activated caspase-9 in living cells
  • Assay utilizes the caspase-9 inhibitor, LEHD-FMK, conjugated to FITC
  • FITC-LEHD-FMK is cell permeable, nontoxic, and irreversibly binds to activated caspase-9 in apoptotic cells
  • FITC label allows detection of activated caspases in apoptotic cells directly by fluorescence microscopy, flow cytometry, or fluorescence plate reader
  • Kit has been configured for research use only and is not for diagnostic and clinical use
  • Store kit at -20°C
Specifications
Specifications
Caspase-9 Staining Kit (Green) is used for sensitive detection of activated caspase-9 in living cells with fluorescein.
Liquid (DMSO)
FITC-LEHD-FMK
Store the kit at -20°C.
1 Kit
Fluorometric
Flow Cytometry, Functional Study