Learn More
Abnova™ Caspase-9 Staining Kit (Red)
Caspase-9 Staining Kit (Red) is used for sensitive detection of activated caspase-9 in living cells with sulfo-rhodamine.
Manufacturer:  Abnova™ KA0765
Catalog No. 89-028-015
Add to cart
Description
Description
  • Kit provides a convenient means for detecting activated caspase-9 in living cells
  • Assay utilizes the caspase-9 inhibitor, LEHD-FMK, conjugated to sulforhodamine (Red-LEHD-FMK) as the fluorescent marker
  • Red-LEHD-FMK is cell permeable, nontoxic, and irreversibly binds to activated caspase-9 in apoptotic cells
  • The fluorescent label allows detection of activated caspase-9 in apoptotic cells directly by fluorescence microscopy, flow cytometry, or fluorescence plate reader
  • Store kit at -20°C
Specifications
Specifications
Caspase-9 Staining Kit (Red) is used for sensitive detection of activated caspase-9 in living cells with sulfo-rhodamine.
Liquid (DMSO)
FITC-LEHD-FMK
Store the kit at -20°C.
1 Kit
Fluorometric
Flow Cytometry, Functional Study