Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Caspase Screening Kit

Caspase Screening Kit is used for detecting activation of caspases by flow cytometry in living cells.

Manufacturer:  Abnova™ KA0724

Catalog No. 89-027-080


Add to cart

Description

Description

  • Kit provides a convenient means for detecting activation of caspases by flow cytometry in living cells
  • Assay is based on the cleavage of (aspartyl) 2-Rhodamine 110 (D2R), a reported substrate for members of caspase family proteases
  • Kit has been configured for research use only and is not for diagnostic and clinical use
  • Kit should be stored at -20°C (store incubation buffer at 4°C after opening)
  • All reagents are stable for 1 year under proper storage conditions

Specifications

Specifications

Caspase Screening Kit is used for detecting activation of caspases by flow cytometry in living cells.
Liquid (DTT), Liquid (DMSO)
1 Year
1 Kit
Functional Study
Store the kit at -20°C.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit