Learn More
Abnova™ Caspase Staining Kit (Red)
Caspase Staining Kit (Red) is used for sensitive detection of activated caspases in living cells with sulfo-rhodamine.
Manufacturer:  Abnova™ KA0726
Catalog No. 89-027-082
Add to cart
Description
Description
  • Kit provides a convenient means for detecting activated caspases in living cells
  • Assay utilizes the caspase family inhibitor VAD-FMK conjugated to sulforhodamine (Red-VAD-FMK) as the fluorescent marker
  • Red-VAD-FMK is cell permeable, nontoxic, and irreversibly binds to activated caspases in apoptotic cells
  • Red fluorescence label allows for direct detection of the activated caspases in apoptotic cells by fluorescence microscopy, flow cytometry, or fluorescence plate reader
  • Kit has been configured for research use only and is not for diagnostic and clinical use
  • Store kit at -20°C
Specifications
Specifications
Caspase Staining Kit (Red) is used for sensitive detection of activated caspases in living cells with sulfo-rhodamine.
Liquid
FITC-VAD-FMK
Store the kit at -20°C.
1 Kit
Fluorometric
Flow Cytometry, Functional Study