Learn More
Abnova™ Caspase Substrate Assay Kit Plus (Colorimetric)
Supplier:  Abnova™ KA3700
 View more versions of this product
Catalog No. 89-101-219
Add to Cart
Description
Description
Specifications
Specifications
Store the kit at -20°C.
Colorimetric
1 Kit
Caspase Substrate Assay Kit (Colorimetric) is used for assaying activities of members of caspase 1-10.
Functional Study
Cell Culture
Product Suggestions
Product Suggestions