Learn More
Abnova™ Catalase Assay Kit
Catalase Assay Kit is used for measuring catalase activity.
Manufacturer:  Abnova™ KA0884
Catalog No. 89-024-439
Add to cart
Description
Description
  • Kit provides a highly sensitive, simple, direct, and HTS-ready assay for measuring Catalase activity in biological samples
  • In the assay, catalase first reacts with hydrogen peroxide to produce water and oxygen
  • Product can be measured using colorimetric or fluorometric methods
  • Catalase activity is reversely proportional to the signal; kit can detect 1 µU or less of catalase activity in samples
  • Store kit at 4°C, protect from warm light
Specifications
Specifications
Colorimetric, Fluorometric
Functional Study
1 Kit
Store the kit at 4°C.