Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cathepsin D Activity Assay Kit

Cathepsin D Activity Assay Kit is use for measuring cathepsin D activity using fluorometric methods.

Manufacturer:  Abnova™ KA0767

Catalog No. 89-028-013


Add to cart

Includes:
  • CD cell lysis buffer, 25mL
  • CD reaction buffer, 5mL
  • CD substrate (1mM), 0.2mL

Description

Description

  • Kit is a fluorescence-based assay that utilizes the preferred cathepsin-D substrate sequence GKPILFFRLK(Dnp)-D-R-NH2 labeled with MCA
  • Released fluorescence can easily be quantified using a fluorometer or fluorescence plate reader
  • Simple, straightforward, and can be adapted to 96-well plate assays
  • Assay conditions have been optimized to obtain the maximal activity
  • Store kit at -20°C (store CD Cell Lysis Buffer and CD Reaction Buffer at 4°C after opening)
  • All reagents are stable for 6 months under proper storage conditions

Specifications

Specifications

Cathepsin D Activity Assay Kit is use for measuring cathepsin D activity using fluorometric methods.
Liquid (Acetic Acid ), Liquid (DMSO)
Functional Study
1 Kit
Fluorometric
Store the kit at -20°C.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit