Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cathepsin G Ab ELISA Kit

indirect solid phase ELISA for the quantitative measurement of IgG class autoantibodies against Cathepsin G in Human Serum or Plasma

Manufacturer:  Abnova™ KA1265

Catalog No. 89-024-557


Add to cart

Specifications

Specifications

HRP
Anti-Cathepsin G Ab ELISA Kit is an indirect solid phase enzyme immunoassay for the quantitative measurement of IgG class autoantibodies against Cathepsin G in human serum or plasma.
Colorimetric
1.5mL Combined calibrator with IgG class anti Cathepsin G 6 vials, 1.5mL Each anti Cathepsin G controls 2 vials, 20mL Sample Buffer 1 vial, 15mL, Enzyme Conjugate solution 1 vial,15mL TMB Substrate solution 1 vial,96 well Plate 1 plate,15mL stop solution
Cathepsin G Ab ELISA kit
Plasma, Serum
100μL
Human
0.5U/mL
Quantification
Cathepsin G Ab ELISA Kit
1 Kit
Store the kit at 4°C.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit