Learn More
Abnova™ Cathepsin K Activity Assay Kit
Cathepsin K Activity Assay Kit is use for measuring cathepsin K activity using fluorometric methods.
Supplier:  Abnova™ KA0769
Catalog No. 89-028-011
Add to Cart
Description
Description
  • Kit is a fluorescence-based assay that utilizes the preferred cathepsin-K substrate sequence LR labeled with AFC
  • Released AFC can easily be quantified using a fluorometer or fluorescence plate reader
  • Simple, straightforward, and can be adapted to 96-well plate assays
  • Assay conditions have been optimized to obtain the maximal activity
  • Store kit at -20°C (store CK Cell Lysis Buffer and CK Reaction Buffer at 4°C after opening)
  • All reagents are stable for 6 months under proper storage conditions
Specifications
Specifications
Cathepsin K Activity Assay Kit is use for measuring cathepsin K activity using fluorometric methods.
1 Kit
Store the kit at -20°C
Functional Study
Product Suggestions
Product Suggestions