Learn More
Abnova™ CCL2 (Human) ELISA Kit Available on GSA/VA Contract for Federal Government customers only.
Sandwich Enzyme Immunoassay for the quantitative measurement of Human CCL2
Manufacturer:  Abnova Corporation KA0169
 View more versions of this product
Catalog No. 89-022-650
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
15.6 to 1000pg/mL
Human
Colorimetric
96-well Antibody coated microwell, 100μL biotin Conjugate Antibody, 150μL Streptavidin HRP, 2 vials standard (lyophilized), 12mL sample diluent, 5mL 20X Assay Buffer concentrate, 50mL Wash Buffer 20X concentrate, 15mL S
96 Assays
4°C
HRP
2.3pg/mL
Quantitative detection of Human CCL2
CCL2 (Human) ELISA kit
Amniotic Fluid, Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
20μL