Learn More
Abnova™ cGMP Direct Immunoassay Kit
cGMP Direct Immunoassay Kit is used for determing cGMP level.
Manufacturer:  Abnova™ KA0887
Catalog No. 89-024-442
Add to cart
Description
Description
  • Kit provides a direct competitive immunoassay for sensitive and quantitative determination of cGMP level in biological samples
  • Kit utilizes the recombinant Protein G coated plate to anchor cGMP polyclonal antibody
  • Provides a new acetylation procedure that improves detection signal significantly
  • Kit can detect 0.04 to 10 pmol/well cGMP samples
  • Store kit at -20°C (after reconstitution, some components may be stored at 4°C as instructed above)
  • Stable for up to 1 to 2 months
Specifications
Specifications
1 Kit
Store the kit at -20°C.
Functional Study
cGMP Direct Immunoassay Kit is used for determing cGMP level.
Colorimetric