Learn More
Abnova™ Chk 2 Gene Knockdown Quantification Kit
Chk 2 Gene Knockdown Quantification Kit is used for measuring Chk2 knockdown induced by siRNA or antisense oligonucleotide at the protein level.
Manufacturer:  Abnova™ KA1544
Catalog No. 89-028-238
Add to cart
Description
Description
  • Quick and efficient procedure, completion of entire assay needs only 4 hours
  • Innovative colorimetric assay without the need for radioactivity, electrophoresis, or chromatography
  • Internal control is conveniently included to correct for the variations for the cell number or protein concentrations
  • Strip microplate format makes the assay flexible, manual or high throughput
  • Simple, reliable, and consistent assay conditions
  • Kit is shipped in two parts: one part at ambient room temperature and the second part on frozen ice packs at 4°C
  • Stable for up to 6 months when stored properly
Specifications
Specifications
Chk 2 Gene Knockdown Quantification Kit is used for measuring Chk2 knockdown induced by siRNA or antisense oligonucleotide at the protein level.
Each
Protein assays and Amino Acid analysis
Store the detection antibody at -20°C. Store Q2, Q4, Q6, GAPDH control antibody and 8-well assay strips at 4°C away from light. Store all other components at room temperature.
Chk 2 Gene knockdown quantification kit
Quantification
10μL