Learn More
Abnova™ Chlamydia pneumoniae IgA ELISA Kit
Determination of IgA autoantibodies against Chlamydia pneumoniae
Manufacturer:  Abnova™ KA1464
Catalog No. 89-028-304
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
HRP
Colorimetric
96-well ELISA break away strip coated with specific Antigen, 1.2mL negative control, 1.2mL positive control, 1.2mL cut off control, 12mL Antigen or Px Conjugate,, 80mL Wash Buffer concentrate,100mL Dilution Buffer,13mL chromogenic Substrate,15mL stop solution
Chlamydia pneumoniae IgA ELISA kit
Serum
100μL
Chlamydia pneumoniae IgA ELISA Kit is a solid phase enzyme immunoassay for the determination of IgA autoantibodies against Chlamydia pneumoniae.
Qualitative
Chlamydia pneumoniae IgA ELISA Kit
1 Kit
Store at 4°C on dry atmosphere away from light.